Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytidine 5'-triphosphate | 65-47-4 | MFCD00047493 | 98.0% | 483.16 | C9H16N3O14P3 | 500MG
SDP

Catalog No. 5000236605
Encompass_Preferred

Cytidine 5'-triphosphate (CTP) is a pyrimidine nucleoside triphosphate supplied as a solid reagent for biochemical research. It functions as a nucleotide building block and substrate for enzymatic reactions, and is commonly used in nucleotide synthesis, enzymatic assays, and lipid biosynthesis studies.

  • Chemical formula C9H16N3O14P3 and molecular weight 483.16.
  • High purity: 98.0%.
  • Appearance: solid.
  • Available in multiple pack sizes, including 500 mg.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Suitable for enzymatic assays and nucleotide synthesis applications.

Catalog No. 50-002-36605 Supplier Medchemexpress LLC Supplier No. HY125818500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytidine 5'-triphosphate (CTP) is a pyrimidine nucleoside triphosphate supplied as a solid reagent for biochemical research. It functions as a nucleotide building block and substrate for enzymatic reactions, and is commonly used in nucleotide synthesis, enzymatic assays, and lipid biosynthesis studies.

  • Chemical formula C9H16N3O14P3 and molecular weight 483.16.
  • High purity: 98.0%.
  • Appearance: solid.
  • Available in multiple pack sizes, including 500 mg.
  • Datasheet, certificate of analysis, and safety data sheet available.
  • Suitable for enzymatic assays and nucleotide synthesis applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.