Learn More
Medchemexpress LLC Cytidine 5'-triphosphate | 65-47-4 | MFCD00047493 | 98.0% | 483.16 g/mol | C9H16N3O14P3 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12581810MM 1ML
Encompass_Preferred
Cytidine-5'-triphosphate is a nucleotide used as a biochemical reagent and substrate in molecular biology and enzymatic assays. Supplied as a ready-to-use 10 mM aqueous solution in a 1 mL vial, it is also available in solid form for larger-scale applications. The compound has a molecular formula of C9H16N3O14P3, a molecular weight of 483.16 g/mol, and a reported purity of 98.0%.
  • Ready-to-use 10 mM aqueous solution in a 1 mL vial.
  • Also available as solid for bulk use.
  • High purity suitable for biochemical assays.
  • Functions as a nucleotide building block for nucleic acid and lipid biosynthesis studies.
  • Soluble in water for direct use in enzymatic reactions.
Catalog No. 50-002-35393