Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytidine 5'-triphosphate | 65-47-4 | MFCD00047493 | 98.0% | 483.16 g·mol⁻¹ | C9H16N3O14P3 | 250MG
SDP

Catalog No. 5000238008
Encompass_Preferred

Cytidine 5'-triphosphate (CTP) is a nucleoside triphosphate used as a biochemical building block for nucleic acid and phospholipid biosynthesis and as a substrate in enzymatic and molecular biology assays. Supplied as a solid with high aqueous solubility, it is suitable for nucleotide incorporation studies, enzyme assays, and general biochemical research.

  • High purity (98.0%).
  • Molecular weight 483.16 g·mol⁻¹.
  • Highly soluble in water (≥200 mg/mL); ultrasonic assistance may be required.
  • Recommended storage: -20°C sealed; in solution -80°C for long-term storage.
  • Available in multiple pack sizes, including 250 MG.

Catalog No. 50-002-38008 Supplier Medchemexpress LLC Supplier No. HY125818250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytidine 5'-triphosphate (CTP) is a nucleoside triphosphate used as a biochemical building block for nucleic acid and phospholipid biosynthesis and as a substrate in enzymatic and molecular biology assays. Supplied as a solid with high aqueous solubility, it is suitable for nucleotide incorporation studies, enzyme assays, and general biochemical research.

  • High purity (98.0%).
  • Molecular weight 483.16 g·mol⁻¹.
  • Highly soluble in water (≥200 mg/mL); ultrasonic assistance may be required.
  • Recommended storage: -20°C sealed; in solution -80°C for long-term storage.
  • Available in multiple pack sizes, including 250 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.