Learn More
Medchemexpress LLC Cytidine-5'-triphosphate-d14 disodium | 00-00-0 | MFCD00078193 | 98.0% | 541.21 g/mol | C9D14N3Na2O14P3 | 5 MG
SDP
Catalog No. 5000264432
Encompass_Preferred
Cytidine-5'-triphosphate-d14 disodium is a deuterium-labeled nucleoside triphosphate provided as the disodium salt for laboratory research. The d14 isotopic label enables sensitive detection by mass spectrometry and NMR and facilitates metabolic tracing and enzymatic synthesis studies. The material is supplied in small, concentrated quantities intended for reconstitution or direct use in analytical workflows.
  • Deuterium-labeled (d14) cytidine triphosphate for isotope tracing.
  • Provided as disodium salt for aqueous solubility.
  • High purity (98.0%) suitable for analytical applications.
  • Supplied in concentrated format for reconstitution or direct use.
  • Useful for mass spectrometry, NMR, and enzymatic synthesis.
Catalog No. 50-002-64432 Supplier Medchemexpress LLC Supplier No. HY125818S15MG
Add to Cart
Add to Cart
Cytidine-5'-triphosphate-d14 disodium is a deuterium-labeled nucleoside triphosphate provided as the disodium salt for laboratory research. The d14 isotopic label enables sensitive detection by mass spectrometry and NMR and facilitates metabolic tracing and enzymatic synthesis studies. The material is supplied in small, concentrated quantities intended for reconstitution or direct use in analytical workflows.
  • Deuterium-labeled (d14) cytidine triphosphate for isotope tracing.
  • Provided as disodium salt for aqueous solubility.
  • High purity (98.0%) suitable for analytical applications.
  • Supplied in concentrated format for reconstitution or direct use.
  • Useful for mass spectrometry, NMR, and enzymatic synthesis.