Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cytidine-5'-triphosphate disodium | 36051-68-0 | 98.0% | 527.12 | 100 MG
SDP

Catalog No. 5000454356
Encompass_Preferred

Cytidine-5'-triphosphate disodium is a nucleoside triphosphate involved in the biosynthesis of DNA, RNA, and lipid. Cytidine triphosphate synthase can catalyze the formation of cytidine 5′-triphosphate from uridine 5′-triphosphate (UTP). This biomolecule is essential in the de novo pyrimidine biosynthetic pathway in *T. gondii* and can regulate cardiolipin biosynthesis in H9c2 cardiac myoblasts.

  • Molecular weight: 527.12
  • Formula: C9H14N3Na2O14P3
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: -20°C, sealed, away from moisture

Catalog No. 50-004-54356 Supplier Medchemexpress LLC Supplier No. HYW013100100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytidine-5'-triphosphate disodium is a nucleoside triphosphate involved in the biosynthesis of DNA, RNA, and lipid. Cytidine triphosphate synthase can catalyze the formation of cytidine 5′-triphosphate from uridine 5′-triphosphate (UTP). This biomolecule is essential in the de novo pyrimidine biosynthetic pathway in *T. gondii* and can regulate cardiolipin biosynthesis in H9c2 cardiac myoblasts.

  • Molecular weight: 527.12
  • Formula: C9H14N3Na2O14P3
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US
  • Storage: -20°C, sealed, away from moisture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.