Learn More
Medchemexpress LLC Cytidine-5'-triphosphate disodium dihydrate | 81012-87-5 | 99.86% | 563.15 | 25 G
SDP
Catalog No. 5000366706
Encompass_Preferred
Cytidine-5'-triphosphate disodium dihydrate (5'-CTP disodium dihydrate) is a high-energy molecule that acts as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation.
  • Acts as a coenzyme in glycerophospholipid biosynthesis.
  • Involved in protein glycosylation.
Catalog No. 50-003-66706 Supplier Medchemexpress LLC Supplier No. 25GHYW013707
Cytidine-5'-triphosphate disodium dihydrate (5'-CTP disodium dihydrate) is a high-energy molecule that acts as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation.
  • Acts as a coenzyme in glycerophospholipid biosynthesis.
  • Involved in protein glycosylation.