Learn More
Medchemexpress LLC Cytidine-5'-triphosphate (disodium dihydrate) | 81012-87-5 | 99.9% | 563.15 | 1 G
SDP
Catalog No. 5000454717
Encompass_Preferred
Cytidine-5'-triphosphate disodium dihydrate (5'-CTP disodium dihydrate) is a high-energy molecule that functions as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation. It is for research use only.
  • Functions as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation
  • Endogenous metabolite
  • Inhibitor of P2X4 purinergic receptor
Catalog No. 50-004-54717 Supplier Medchemexpress LLC Supplier No. HYW0137071G
Cytidine-5'-triphosphate disodium dihydrate (5'-CTP disodium dihydrate) is a high-energy molecule that functions as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation. It is for research use only.
  • Functions as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation
  • Endogenous metabolite
  • Inhibitor of P2X4 purinergic receptor