Learn More
Medchemexpress LLC Cytidine-5'-triphosphate (disodium dihydrate) | 81012-87-5 | 99.9% | 563.15 | 10 G
SDP
Catalog No. 5000454143
Encompass_Preferred
Cytidine-5'-triphosphate disodium dihydrate (5'-CTP disodium dihydrate) is a high-energy molecule that functions as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation.
  • Acts as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation.
Catalog No. 50-004-54143 Supplier Medchemexpress LLC Supplier No. HYW01370710G
Cytidine-5'-triphosphate disodium dihydrate (5'-CTP disodium dihydrate) is a high-energy molecule that functions as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation.
  • Acts as a coenzyme in glycerophospholipid biosynthesis and protein glycosylation.