Learn More
Medchemexpress LLC Cytidine 5'-triphosphate disodium salt | 36051-68-0 | MFCD00078193 | >97.0% | 527.12 g/mol | C9H14N3Na2O14P3 | 1ML
SDP
Catalog No. 5000236526
Encompass_Preferred
CTP disodium salt is a ready-to-use, PCR-grade 100 mM solution of cytidine-5'-triphosphate (disodium salt) supplied in a 1 mL vial. The formulation is qualified for molecular biology use and is intended as a nucleotide substrate for enzymatic reactions and nucleic acid synthesis.
Catalog No. 50-002-36526 Supplier Medchemexpress LLC Supplier No. HY125818I1ML
CTP disodium salt is a ready-to-use, PCR-grade 100 mM solution of cytidine-5'-triphosphate (disodium salt) supplied in a 1 mL vial. The formulation is qualified for molecular biology use and is intended as a nucleotide substrate for enzymatic reactions and nucleic acid synthesis.