Learn More
Alkali Scientific Cytidine, 500 G
SDP
Catalog No. 5000663240
Encompass_Preferred
Cytidine is a pyrimidine nucleoside consisting of cytosine linked to a ribose sugar and is a key building block of RNA molecules. It plays a vital role in nucleic acid metabolism, RNA synthesis, and cellular biochemical processes. In molecular biology research, cytidine is frequently used in studies examining RNA transcription, nucleotide metabolism, and enzymatic reactions involving nucleosides. Scientists utilize cytidine in biochemical assays to investigate nucleic acid synthesis, cellular regulation, and metabolic pathways related to nucleotide turnover. Because of its importance in RNA structure and function, cytidine is widely used in life science laboratories for nucleic acid research, molecular biology experiments, and biochemical pathway analysis.
Catalog No. 50-006-63240 Supplier Alkali Scientific Supplier No. YTI003PL
Cytidine is a pyrimidine nucleoside consisting of cytosine linked to a ribose sugar and is a key building block of RNA molecules. It plays a vital role in nucleic acid metabolism, RNA synthesis, and cellular biochemical processes. In molecular biology research, cytidine is frequently used in studies examining RNA transcription, nucleotide metabolism, and enzymatic reactions involving nucleosides. Scientists utilize cytidine in biochemical assays to investigate nucleic acid synthesis, cellular regulation, and metabolic pathways related to nucleotide turnover. Because of its importance in RNA structure and function, cytidine is widely used in life science laboratories for nucleic acid research, molecular biology experiments, and biochemical pathway analysis.