Learn More
Medchemexpress LLC HY-B0158 500mg Medchemexpress, Cytidine CAS:65-46-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0158500MG
Encompass_Preferred
Medchemexpress, HY-B0158 500mg Cytidine CAS:65-46-3 Cytidine is a pyrimidine nucleoside and acts as a component of RNA. Cytidine is a precursor of uridine. Cytidine controls neuronal-glial glutamate cycling, affecting cerebral phospholipid metabolism, catecholamine synthesis, and mitochondrial function. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2384