Learn More
Apexbio Technology LLC Cytidine 65-46-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089939
Encompass_Preferred
Cytidine (CAS 65-46-3) is a pyrimidine nucleoside composed of cytosine linked to a ribofuranose moiety As an integral constituent of RNA cytidine participates in the synthesis of nucleic acids and mediates genetic information transfer through base pairing during transcription It also plays roles in cellular metabolism and the modulation of epigenetic processes via incorporation into RNA Cytidine is frequently utilized in research to investigate RNA structure and function nucleotide metabolism and as a substrate in enzymatic assays involving nucleoside transporters kinases or deaminases
Catalog No. 50-000-89939 Supplier Apexbio Technology LLC Supplier No. B19235.1
Cytidine (CAS 65-46-3) is a pyrimidine nucleoside composed of cytosine linked to a ribofuranose moiety As an integral constituent of RNA cytidine participates in the synthesis of nucleic acids and mediates genetic information transfer through base pairing during transcription It also plays roles in cellular metabolism and the modulation of epigenetic processes via incorporation into RNA Cytidine is frequently utilized in research to investigate RNA structure and function nucleotide metabolism and as a substrate in enzymatic assays involving nucleoside transporters kinases or deaminases