Learn More
Medchemexpress LLC Cytidine (standard) | 65-46-3 | 99.9% | 1 ML
SDP
Catalog No. 5000406942
Encompass_Preferred
Cytidine is a pyrimidine nucleoside that functions as a component of RNA and is a precursor of uridine. It plays a role in controlling neuronal-glial glutamate cycling, which impacts cerebral phospholipid metabolism, catecholamine synthesis, and mitochondrial function.
  • Functions as a component of RNA
  • Precursor of uridine
  • Regulates neuronal-glial glutamate cycling
  • Influences cerebral phospholipid metabolism, catecholamine synthesis, and mitochondrial function
  • Demonstrates inhibitory effects on various cancer cell lines
  • Reduces glutamate/glutamine levels in vivo
  • May improve depressive symptoms
Catalog No. 50-004-06942 Supplier Medchemexpress LLC Supplier No. HYB015810MM/1ML
Cytidine is a pyrimidine nucleoside that functions as a component of RNA and is a precursor of uridine. It plays a role in controlling neuronal-glial glutamate cycling, which impacts cerebral phospholipid metabolism, catecholamine synthesis, and mitochondrial function.
  • Functions as a component of RNA
  • Precursor of uridine
  • Regulates neuronal-glial glutamate cycling
  • Influences cerebral phospholipid metabolism, catecholamine synthesis, and mitochondrial function
  • Demonstrates inhibitory effects on various cancer cell lines
  • Reduces glutamate/glutamine levels in vivo
  • May improve depressive symptoms