Learn More
Aviva Systems Biology Corp CYTIP Antibody - N-terminal region (ARP62400_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62400_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CYTIP. It was validated on Western Blot. . Alternate names: B3-1, CASP, CYBR, CYTHIP, HE, PSCDBPApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Pig, Rat
Catalog No. 50-287-670