Learn More
AdipoGen Cytisine
SDP
Supplier:  AdipoGen CDXC0228M100
Encompass_Preferred
Chemical. CAS 485-35-8. Formula C11H14N2O. MW 190.2. Synthetic. Pyridine-like alkaloid. Nicotine agonist. Potent agonist at alpha3beta4 and alpha7 nicotinic acetylcholine receptors and partial agonist at alpha4beta2 nicotinic acetylcholine receptors. Shows analgesic, antihypertensive and inotropic activities. Used for smoking cessation. Iron-chelator and hydroxyl radical scavanger. Has antidepressant properties. Used in forensics in novel surface molecular imprinting techniques.
Catalog No. 50-256-1427