Learn More
Medchemexpress LLC HY-N0175 25mg Medchemexpress, Cytisinicline CAS:485-35-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN017525MG
Encompass_Preferred
Medchemexpress, HY-N0175 25mg Cytisinicline CAS:485-35-8 Cytisinicline (Cytisine) is an alkaloid that occurs naturally in several plant genera, such as Laburnum and Cytisus. Cytisinicline (Cytisine) is a partial agonist of α4β2 nAChRs [1] , and partial to full agonist at β4 containing receptors and α7 receptors [2] . Has been used medically to help with smoking cessation [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3446