Learn More
Genetex Inc Cytokeratin 6 Monoclonal Antibody [MSVA-606R] HistoMAX™ (GTX04403) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04403
Encompass_Preferred
The Cytokeratin 6 antibody [MSVA-606R] HistoMAX™ (GTX04403) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human colorectal adenocarcinoma (COAD) tissue using GTX04403 Cytokeratin 6 antibody [MSVA-606R] HistoMAX. Colorectal adenocarcinoma with strong KRT6 positivity in about 50% of tumor cells.
Catalog No. NC3666770