Learn More
Genetex Inc Cytokeratin 7 Monoclonal Antibody [MSVA-607R] HistoMAX™ (GTX04375) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04375
Encompass_Preferred
The Cytokeratin 7 antibody [MSVA-607R] HistoMAX™ (GTX04375) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human fallopian tube tissue using GTX04375 Cytokeratin 7 antibody [MSVA-607R] HistoMAX. All epithelial cells but ciliated cells stain cytokeratin 7 positive.
Catalog No. NC3665626