Learn More
Invitrogen™ CytKick™ Max Autosampler, refurbished
Catalog No. A42973R
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 each
Catalog No. Quantity
A42973R 1 each
1 options
Catalog No. A42973R Supplier Invitrogen™ Supplier No. A42973R
Please contact your sales representative for a quote or further ordering instructions.
The CytKick Max Autosampler Refurbished is for use with the Attune NxT Acoustic Focusing Cytometer. It enables rapid processing of multiple samples by automatically analyzing samples from 96- and 384-well plates (standard and deep), as well as some custom plate types and microfuge tube racks.
This is a refurbished version of the CytKick Max Autosampler, which is for use with the Attune NxT Acoustic Focusing Cytometer. It enables rapid processing of multiple samples by automatically analyzing samples from 96- and 384-well plates (standard and deep), as well as some custom plate types and microfuge tube racks.
TRUSTED_SUSTAINABILITY
Specifications
Product Line CytKick
Quantity 1 each
Product Type Flow Cytometry Analyzer