Learn More
Enzo Life Sciences CYTO-ID Autophagy detection kit (1x50tests)
SDP
Catalog No. 501031812
Encompass_Preferred
A no-transfection, quantitative assay for monitoring autophagy in live cells. CYTO-ID.The probe is a cationic amphiphilic tracer (CAT) dye that rapidly partitions into cells in a similar manner as drugs that induce phospholipidosis. Careful selection of titratable functional moieties on the dye prevents its accumulation within lysosomes, but enables labeling of vacuoles associated with the autophagy pathway. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection, HTS. Contents: CYTO-ID Green Detection Reagent. Hoechst 33342 Nuclear Stain. Autophagy Inducer (Rapamycin). Chloroquine Control. 10X Assay Buffer
Catalog No. 50-103-1812 Supplier Enzo Life Sciences Supplier No. ENZ510310050
A no-transfection, quantitative assay for monitoring autophagy in live cells. CYTO-ID.The probe is a cationic amphiphilic tracer (CAT) dye that rapidly partitions into cells in a similar manner as drugs that induce phospholipidosis. Careful selection of titratable functional moieties on the dye prevents its accumulation within lysosomes, but enables labeling of vacuoles associated with the autophagy pathway. Applications: Flow Cytometry. Fluorescence microscopy. Fluorescent detection, HTS. Contents: CYTO-ID Green Detection Reagent. Hoechst 33342 Nuclear Stain. Autophagy Inducer (Rapamycin). Chloroquine Control. 10X Assay Buffer