Learn More
Description
Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.
- Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
- Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
- Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
- High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
- Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
- Emission can be collected in either the PE-Cy5 or APC channels
- Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters
Recommended for:
- Open platform microspheres for suspension array analysis in flow cytometry
Specifications
Specifications
| Additive | Sodium Azide Preservative, 0.05% Tween-20 |
| Content And Storage | Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer. |
| Contents | Polystyrene microspheres in water |
| Description | Cyto-Plex Carboxylated Particles, 4µm |
| Color | Red |
| Material | Polystyrene with Copolymer Grafted Surface |
| Product Type | Carboxylated Particle |
| Dye Type | Firefli™ Fluorescent Red (635/700nm) |
| Includes | Material Safety Data Sheet (MSDS) and Package Insert Sheet |
| Level | Level 4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.