Learn More
Electron Microscopy Sciences CytoPrep™ Fixative 4 OZ
SDP
Supplier:  Electron Microscopy Sciences HY3272125MG
Encompass_Preferred
CytoPrep Fixative forms a clear protective coating over specimens while fixing cells. It is ideal for cytological smears and it dries within 4 to 6 minutes. Good for safe storage and transportation of slides. It is water soluble and contains no chlorofluorocarbons.
Catalog No. 50-252-70