Learn More
Electron Microscopy Sciences Cyto-Slide Vce & Lines
SDP
Supplier:  Electron Microscopy Sciences 7187401
Encompass_Preferred
Special glass slides measuring 1x3" (25x75 mm) and 1 mm thick. Superfrost letters VCE and lines on the same side as the superfrost end.
Catalog No. 50-949-353