Learn More
LabScientific Cyto-spin, uncoated microscope slides. 72/pack. Precise sample framing. Permanent hydrophobic epoxy ink resistant to solvents.
Catalog No. NC1095425
Encompass
Cyto-Spin uncoated slides to locate centrifuge-smears quickly by precisely framing sample. 72/pack. Precise sample framing. Permanent hydrophobic epoxy ink resistant to solvents.
Catalog No. NC1095425 Supplier LabScientific Supplier No. 0621
Cyto-Spin uncoated slides to locate centrifuge-smears quickly by precisely framing sample. 72/pack. Precise sample framing. Permanent hydrophobic epoxy ink resistant to solvents.