Learn More
Electron Microscopy Sciences Cyto-Tek 6 ml Fluid Chamber
SDP
Catalog No. 5025281
Encompass
Enhance your cell recovery up to 100%. The funnel chanber has highly polished specimen chamber that minimizes cell attachment and a 33% thicker clamp wall (for greater claming tension) that provides paced fluid absorption. The window ledge to helps secure filter and keep cells in target area. A "V" cutout adds rigidity helping to hold funnel chamber in place.
Catalog No. 50-252-81 Supplier Electron Microscopy Sciences Supplier No. 6261206
Enhance your cell recovery up to 100%. The funnel chanber has highly polished specimen chamber that minimizes cell attachment and a 33% thicker clamp wall (for greater claming tension) that provides paced fluid absorption. The window ledge to helps secure filter and keep cells in target area. A "V" cutout adds rigidity helping to hold funnel chamber in place.