Learn More
Coopersurgical Inc CYTOBRUSH + CELL COLL 100/PK
Catalog No. NC9458596
Encompass_Preferred
Cytobrush Plus - 10 Bags, 10 Brushes each (100/Box). The classic Cytobrush Plus Cell collector for endocervical sampling is excellent for nulliparous or menopausal women, or for collecting endocervical cells at conization. The gentle brush collects large amounts of cells that are easily transferred to slides for analysis.
Catalog No. NC9458596 Supplier Coopersurgical Inc Supplier No. C0004
Cytobrush Plus - 10 Bags, 10 Brushes each (100/Box). The classic Cytobrush Plus Cell collector for endocervical sampling is excellent for nulliparous or menopausal women, or for collecting endocervical cells at conization. The gentle brush collects large amounts of cells that are easily transferred to slides for analysis.