Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ CytoCell Block Fixative
SDP

Catalog No. STLCBF32
Change view
Click to view available options
Product Type:
Fixative
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
STLCBF32 Fixative
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.
Catalog No. STLCBF32 Supplier StatLab™ Supplier No. CBF32
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, in contact with skin, or inhaled.

CytoCell Block Fix is designed as a fixative for cells from body fluids.

  • Ideal for fine needle aspirations, body cavity fluids, washings, brushings, and gynecologic and non-gynecologic liquid-based specimens
  • CytoCell Block Fix causes precipitation of protein and trapping of cells within the protein matrix present
  • Nuclei show crisp nuclear membranes with distinct nucleoli

Specifications

Color Clear
For Use With (Equipment) All Tissue Processors
Grade Chemical Solution
Product Type Fixative
Quantity 1 L
For Use With (Application) Fixation
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.