Learn More
Cayman Chemical CytochalasIn A 10mg
SDP
Catalog No. 502955265
Encompass_Preferred
Inhibits actin polymerization; uniquely inhibits HIV-1 protease (IC50 = 3 μM); rapidly and reversibly inhibits glucose transport by competitively binding glucose transporters (Ki = 4.0 μM); induces the phosphorylation of the tyrosine phosphatase PTP3 of Dictyostelium, activating STATc
Catalog No. 50-295-5265 Supplier Cayman Chemical Supplier No. 1132710 MG
Inhibits actin polymerization; uniquely inhibits HIV-1 protease (IC50 = 3 μM); rapidly and reversibly inhibits glucose transport by competitively binding glucose transporters (Ki = 4.0 μM); induces the phosphorylation of the tyrosine phosphatase PTP3 of Dictyostelium, activating STATc