Learn More
Cayman Chemical Cytochalasin A, Purity: ≥98%, CAS: 14110-64-6, 1 mg
SDP
Catalog No. 502955585
Encompass_Preferred
Inhibits actin polymerization; uniquely inhibits HIV-1 protease (IC50 = 3 μM); rapidly and reversibly inhibits glucose transport by competitively binding glucose transporters (Ki = 4.0 μM); induces the phosphorylation of the tyrosine phosphatase PTP3 of Dictyostelium, activating STATc
Catalog No. 50-295-5585 Supplier Cayman Chemical Supplier No. 113271 MG
Inhibits actin polymerization; uniquely inhibits HIV-1 protease (IC50 = 3 μM); rapidly and reversibly inhibits glucose transport by competitively binding glucose transporters (Ki = 4.0 μM); induces the phosphorylation of the tyrosine phosphatase PTP3 of Dictyostelium, activating STATc