Learn More
Enzo Life Sciences Cytochalasin A (5mg). CAS: 14110-64-6
SDP
Supplier:  Enzo Life Sciences ALX380057M005
Encompass_Preferred
Antibiotic. Fungal toxin. Inhibits glucose transport, actin polymerisation and blocks the formation of microtubuli. Inhibits cell division. Inhibits HIV-1 protease. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in acetone, DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-8936