Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochalasin A, 99%, MP Biomedicals™

Catalog No. ICN15076901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15076901 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15076901 Supplier MP Biomedicals, Inc Supplier No. 0215076901

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14110-64-6
Molecular Formula C29H35NO5
Molecular Weight (g/mol) 477.60
MDL Number MFCD00005935
InChI Key ZMAODHOXRBLOQO-TZVKRXPSSA-N
Synonym cytochalasin a from drechslera dematioidea, cytochalasin a from helminthosporium dematioideum, 2h-oxacyclotetradecino 2,3-d isoindole-2,5,18-trione,6,7,8,9,10,12a,13,14,15,15a,16,17-dodecahydro-13-hydroxy-9,15-dimethyl-14-methylene-16-phenylmethyl-, 3e,9r,11e,12as,13s,15s,15as,16s,18as
PubChem CID 57431204
IUPAC Name (9R,12aS,13S,15S,15aS,16S,18aS)-16-benzyl-13-hydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,5,18-trione
SMILES C[C@H]1[C@H]2[C@H](CC3=CC=CC=C3)NC(=O)[C@]22OC(=O)\C=C\C(=O)CCC[C@@H](C)C\C=C\[C@H]2[C@H](O)C1=C

Specifications

Color White
Assay Percent Range 0.99
Quantity 1 mg
Formula Weight 477.6
Physical Form Powder
Chemical Name or Material Cytochalasin A, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.