Learn More
Cayman Chemical Cytochalasin B, Purity: ≥98%, CAS: 14930-96-2, 10 mg
SDP
Catalog No. 502955707
Encompass_Preferred
A cell-permeable mycotoxin which binds to the barbed end of actin, reversibly inhibiting the elongation and shortening of actin filaments; blocks diverse cellular functions, including cell division, migration, phagocytosis, exocytosis, chemotaxis, and glucose transport
Catalog No. 50-295-5707 Supplier Cayman Chemical Supplier No. 1132810 MG
A cell-permeable mycotoxin which binds to the barbed end of actin, reversibly inhibiting the elongation and shortening of actin filaments; blocks diverse cellular functions, including cell division, migration, phagocytosis, exocytosis, chemotaxis, and glucose transport