Learn More
Crescent Chemical Co Inc CYTOCHALASIN B
SDP
Supplier:  Crescent Chemical Co Inc SE18015.01
Encompass_Preferred
Cytochalasin B From Helminthosporium dematoideum. Reversible inhibitor of cell motion (1). Inhibits phagocytosis (2). Induces polyploidy (3). Induces nuclear extrusion (4, 5). Size - 1MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 18015.
Catalog No. 50-247-461