Learn More
Medchemexpress LLC Cytochalasin B | 14930-96-2 | 99.6% | 479.61 | 1 ML
SDP
Catalog No. 5000393744
Encompass_Preferred
Cytochalasin B is a cell-permeable mycotoxin that disrupts the formation of actin polymers by binding to the barbed end of actin filaments. It also blocks cell migration and exhibits a Kd value of 1.4-2.2 nM for F-actin.
  • Cell-permeable mycotoxin
  • Binds to barbed end of actin filaments
  • Disrupts actin polymer formation
  • Blocks cell migration
Catalog No. 50-003-93744 Supplier Medchemexpress LLC Supplier No. HY1692810MM/1ML
Cytochalasin B is a cell-permeable mycotoxin that disrupts the formation of actin polymers by binding to the barbed end of actin filaments. It also blocks cell migration and exhibits a Kd value of 1.4-2.2 nM for F-actin.
  • Cell-permeable mycotoxin
  • Binds to barbed end of actin filaments
  • Disrupts actin polymer formation
  • Blocks cell migration