Learn More
AdipoGen Cytochalasin B (5 mg)
SDP
Supplier:  AdipoGen AGCN20504M005
Encompass_Preferred
Chemical. CAS: 14930-96-2. Formula: C29H37NO5. MW: 479.6. Isolated from Phoma sp. Cell permeable mycotoxin. Actin polymerization inhibitor. Binds to the barbed end of actin, reversibly inhibiting the elongation and shortening of actin filaments. Induces nuclear extrusion. By disrupting actin polymerization, blocks diverse cellular functions, including cell division, migration, phagocytosis, exocytosis and chemotaxis. Induces DNA fragmentation. Used in cytoskeletal reorganization, cell imaging and organelle trafficking studies. Potent antitumor activity. Causes cell cycle arrest and induces apoptosis in cancer cells. Shown to have an inhibitory effect on tumor cell growth without causing immunosuppressive effects. Non-competitive glucose transport inhibitor. Inhibits of GLUT1/GLUT2 mediated monosaccharide transport across the plasma membrane. Does not inhibit GLUT5. Used for testing the genotoxicity of substances in the cytokinesis-block micronucleus cytome (CBMN cyt) assay.
Catalog No. 50-205-3687