Learn More
Abcam Cytochalasin B, actin polymerization inhibitor, 5MG
Supplier:  Abcam AB1434825MG
Encompass_Preferred
MW 479.6 Da, Purity >97%. Cell permeable actin polymerization inhibitor. Reversible KV1.5 blocker (IC₅₀ = 4 μM). Inhibits I(Kur). Cytochalasin A (ab143481) analog. Inhibits transport of monosaccharides across the cell membrane. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1280600