Learn More
Crescent Chemical Co Inc CYTOCHALASIN B
SDP
Catalog No. 50247462
Encompass_Preferred
Cytochalasin B From Helminthosporium dematoideum. Reversible inhibitor of cell motion (1). Inhibits phagocytosis (2). Induces polyploidy (3). Induces nuclear extrusion (4, 5). Size - 5MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 18015.
Catalog No. 50-247-462 Supplier Crescent Chemical Co Inc Supplier No. SE18015.02
Cytochalasin B From Helminthosporium dematoideum. Reversible inhibitor of cell motion (1). Inhibits phagocytosis (2). Induces polyploidy (3). Induces nuclear extrusion (4, 5). Size - 5MG Storage Conditions - +2 °C TO +8 °C Catalog Number - 18015.