Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochalasin B, >99%, For HPLC analysis, MP Biomedicals™

Catalog No. ICN19511901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19511901 1 mg
ICN19511905 5 mg
ICN19511910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19511901 Supplier MP Biomedicals, Inc Supplier No. 0219511901
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14930-96-2
Molecular Formula C29H37NO5
Molecular Weight (g/mol) 479.62
MDL Number MFCD00077704
InChI Key GBOGMAARMMDZGR-UHFFFAOYNA-N
Synonym cytochalasin b, cytochalasin b from helminthosporium dematioideum
PubChem CID 44634701
IUPAC Name 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione
SMILES CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C

Specifications

Color White
Quantity 1 mg
Formula Weight 479.6
Percent Purity >99%
Physical Form Powder
Chemical Name or Material Cytochalasin B, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.