Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochalasin B, MP Biomedicals™

Catalog No. MP219511925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219511925 25 mg
MP219511950 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP219511925 Supplier MP Biomedicals, Inc Supplier No. 0219511925
Add to Cart
Add to Cart

Disrupts contractile microfilaments by inhibiting actin polymerization. Cytochalasin B, MP Biomedicals is a cell permeable fungal toxin that is an important tool for cytological research. It induces DNA fragmentation, inhibits cell division, disrupts many

Cytochalasin B is one of a group of fungal metabolites that interfere with a wide variety of cellular movements. It is a useful tool for characterizing some of the polymerization properties of actin, and in studies on cytokinesis. It is used as a probe for the two hexose-transport systems in rat L6 myoblasts.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14930-96-2
Molecular Formula C29H37NO5
Molecular Weight (g/mol) 479.62
MDL Number MFCD00077704
InChI Key GBOGMAARMMDZGR-UHFFFAOYNA-N
Synonym cytochalasin b, cytochalasin b from helminthosporium dematioideum
PubChem CID 44634701
IUPAC Name 16-benzyl-5,13-dihydroxy-9,15-dimethyl-14-methylidene-2H,5H,6H,7H,8H,9H,10H,12aH,13H,14H,15H,15aH,16H,17H,18H-oxacyclotetradeca[3,2-e]isoindole-2,18-dione
SMILES CC1C2C(CC3=CC=CC=C3)NC(=O)C22OC(=O)C=CC(O)CCCC(C)CC=CC2C(O)C1=C

Specifications

Melting Point 216°C to 226°C
Quantity 25 mg
Solubility Information Soluble in methanol (20mg/mL, clear, colorless solution) or ethanol, DMSO (10mg/mL), DMF; Insoluble in water.
Formula Weight 479.6g/mol
Physical Form Powder
Recommended Storage Store at −20°C, desiccate, protect from light.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.