Learn More
Cayman Chemical CytochalasIn C 1mg
SDP
Catalog No. 502953170
Encompass_Preferred
An actin polymerization inhibitor and fungal metabolite; inhibits actin polymerization at 10 µM; cytotoxic to HeLa cells (IC50 = <0.32 UG/ml)
Catalog No. 50-295-3170 Supplier Cayman Chemical Supplier No. 113291 MG
Add to Cart
Add to Cart
An actin polymerization inhibitor and fungal metabolite; inhibits actin polymerization at 10 µM; cytotoxic to HeLa cells (IC50 = <0.32 UG/ml)