Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cell Signaling Technology Cytochalasin D, >98% Purity, 1 mg
SDP

Catalog No. 502729862
Encompass

Cytochalasin D is a fungal-derived toxin that acts as a potent, specific inhibitor of actin polymerization. It limits elongation at both the barbed and pointed ends of actin filaments, making it an effective tool for studying cytoskeleton-mediated processes including signal transduction, protein transport, and cell motility. This product is supplied as a lyophilized powder.

  • Purity: >98%
  • Molecular weight: 507.6 g/mol
  • Molecular formula: C30H37NO6
  • Solubility: Soluble in DMSO (20 mg/mL) or ethanol (5 mg/mL)
  • Reconstitution: For a 5 mM stock, reconstitute 1 mg of powder in 0.39 mL of DMSO
  • Storage: Store lyophilized at -20°C, desiccated. Once in solution, store at -20°C and use within 3 months

Catalog No. 50-272-9862 Supplier Cell Signaling Technology Supplier No. 94946S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cytochalasin D is a fungal-derived toxin that acts as a potent, specific inhibitor of actin polymerization. It limits elongation at both the barbed and pointed ends of actin filaments, making it an effective tool for studying cytoskeleton-mediated processes including signal transduction, protein transport, and cell motility. This product is supplied as a lyophilized powder.

  • Purity: >98%
  • Molecular weight: 507.6 g/mol
  • Molecular formula: C30H37NO6
  • Solubility: Soluble in DMSO (20 mg/mL) or ethanol (5 mg/mL)
  • Reconstitution: For a 5 mM stock, reconstitute 1 mg of powder in 0.39 mL of DMSO
  • Storage: Store lyophilized at -20°C, desiccated. Once in solution, store at -20°C and use within 3 months

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.