Learn More
Cayman Chemical Cytochalasin D, Purity: ≥95%, CAS: 22144-77-0, 10 mg
SDP
Catalog No. 502953693
Encompass_Preferred
A cell-permeable inhibitor that binds actin filaments, but not actin monomers, to inhibit polymerization at concentrations as low as 0.2 µM
Catalog No. 50-295-3693 Supplier Cayman Chemical Supplier No. 1133010 MG
A cell-permeable inhibitor that binds actin filaments, but not actin monomers, to inhibit polymerization at concentrations as low as 0.2 µM