Learn More
Cayman Chemical Cytochalasin D, Purity: ≥95%, CAS: 22144-77-0, 1 mg
SDP
Catalog No. 502953238
Encompass_Preferred
A cell-permeable inhibitor that binds actin filaments, but not actin monomers, to inhibit polymerization at concentrations as low as 0.2 µM
Catalog No. 50-295-3238 Supplier Cayman Chemical Supplier No. 113301 MG
A cell-permeable inhibitor that binds actin filaments, but not actin monomers, to inhibit polymerization at concentrations as low as 0.2 µM