Learn More
Medchemexpress LLC Cytochalasin D | 22144-77-0 | 25 MG
SDP
Catalog No. 5000420549
Encompass_Preferred
Cytochalasin D (Zygosporin A) is a potent, cell-permeable inhibitor of actin polymerization, derived from fungus. It works by binding to G-actin, thereby inhibiting the interaction between G-actin and cofilin. This action also reduces the binding of cofilin to F-actin, consequently decreasing the rate of both actin polymerization and depolymerization in living cells.
  • Effectively disrupts the dynamics of actin filaments.
  • Can easily enter cells to exert its effects.
  • Prevents cofilin from interacting with G-actin.
  • Modulates actin dynamics within cells.
  • Can lower the amount of survivin in the tumor environment by reducing exosome release.
  • Affects the localization and activity of the Yap protein.
Catalog No. 50-004-20549 Supplier Medchemexpress LLC Supplier No. 25MGHYN6682
Cytochalasin D (Zygosporin A) is a potent, cell-permeable inhibitor of actin polymerization, derived from fungus. It works by binding to G-actin, thereby inhibiting the interaction between G-actin and cofilin. This action also reduces the binding of cofilin to F-actin, consequently decreasing the rate of both actin polymerization and depolymerization in living cells.
  • Effectively disrupts the dynamics of actin filaments.
  • Can easily enter cells to exert its effects.
  • Prevents cofilin from interacting with G-actin.
  • Modulates actin dynamics within cells.
  • Can lower the amount of survivin in the tumor environment by reducing exosome release.
  • Affects the localization and activity of the Yap protein.