Learn More
Selleck Chemical LLC Cytochalasin D
SDP
Catalog No. 5000001905
Encompass_Preferred
Cytochalasin D (Zygosporin A NSC 209835) is a potent actin polymerization inhibitor derived from fungus Cytochalasin D inhibits actin filament dynamics in cells It induces phosphorylation and cytoplasmic retention of Yap
Catalog No. 50-000-01905 Supplier Selleck Chemical LLC Supplier No. S81841MG
Add to Cart
Add to Cart
Cytochalasin D (Zygosporin A NSC 209835) is a potent actin polymerization inhibitor derived from fungus Cytochalasin D inhibits actin filament dynamics in cells It induces phosphorylation and cytoplasmic retention of Yap