Learn More
Selleck Chemical LLC Cytochalasin D
SDP
Supplier:  Selleck Chemical LLC S81845MG
Encompass_Preferred
Cytochalasin D (Zygosporin A NSC 209835) is a potent actin polymerization inhibitor derived from fungus Cytochalasin D inhibits actin filament dynamics in cells It induces phosphorylation and cytoplasmic retention of Yap
Catalog No. 50-000-01907