Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytochalasin D, MP Biomedicals™

Catalog No. MP215077110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215077110 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215077110 Supplier MP Biomedicals, Inc Supplier No. 0215077110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disrupts actin filaments and inhibits actin polymerization. Cytochalasin D, MP Biomedicals is a cell-permeable fungal toxin that binds to the barbed end of actin filaments, inhibiting both the association and dissociation of subunits. It possesses antibio

This compound disrupts actin microfilaments and activates the p53-dependent pathways, causing arrest of the cell cycle at the G1-S transition.

  • Inhibits smooth muscle contraction
  • Inhibits insulin-stimulated, but not basal, transport of glucose
  • Possesses antibiotic and antitumor activity
  • Impairs maintenance of long term potentiation (LTP) of actin filaments
  • Implicated in promoting conditions favorable for depolymerizing actin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22144-77-0
Molecular Formula C30H37NO6
Molecular Weight (g/mol) 507.627
InChI Key SDZRWUKZFQQKKV-DKLHZCHASA-N
Synonym cytochalasin d
PubChem CID 131668624
SMILES CC1CC=CC2C(C(=C)C(C3C2(C(C=CC(C1=O)(C)O)OC(=O)C)C(=O)NC3CC4=CC=CC=C4)C)O

Specifications

Melting Point 193°C to 195°C (literature)
Quantity 10 mg
Solubility Information Soluble in chloroform (10g/mL, clear, colorless solution), CH2Cl2 (10mg/ mL, clear colorless solution), DMSO and ethanol.
Formula Weight 507.6
Physical Form Powder
Recommended Storage Store at −20°C, protect from light, dessicated.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.