Learn More
Cayman Chemical Cytochalasin E, Purity: ≥98%, CAS: 36011-19-5, 10 mg
SDP
Catalog No. 502955382
Encompass_Preferred
Inhibits actin polymerization; potently and selectively inhibits the growth of endothelial cells (IC50 1 protease activity
Catalog No. 50-295-5382 Supplier Cayman Chemical Supplier No. 1133110 MG
Inhibits actin polymerization; potently and selectively inhibits the growth of endothelial cells (IC50 1 protease activity