Learn More
Cayman Chemical CytochalasIn E 1mg
SDP
Supplier:  Cayman Chemical 113311 MG
Encompass_Preferred
Inhibits actin polymerization; potently and selectively inhibits the growth of endothelial cells (IC50 1 protease activity
Catalog No. 50-295-3073