Learn More
Cayman Chemical Cytochalasin H, Purity: ≥95%, CAS: 53760-19-3, 1 mg
SDP
Catalog No. 502967675
Encompass_Preferred
A potent inhibitor of actin incorporation into filaments; also increases the steady-state diffusion coefficients of filaments, suggesting that it stimulates filament shortening
Catalog No. 50-296-7675 Supplier Cayman Chemical Supplier No. 184341 MG
A potent inhibitor of actin incorporation into filaments; also increases the steady-state diffusion coefficients of filaments, suggesting that it stimulates filament shortening